Learn More
Restek™ Scoring Wafer with Handle
SDP
Catalog No. 06712122 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Scoring Wafer with Ergonomic Rubber Handle and Ceramic Two-Sided Wafer
Catalog No.
06-712-122 Scoring Wafer with Ergonomic Rubber Handle and Ceramic Two-Sided Wafer
1 options
This item is not returnable. View return policy
Catalog No. 06-712-122 Supplier Restek™ Supplier No. 23015
This item is not returnable. View return policy
Straight edged for cutting fused silica tubing cleanly
  • Cuts both fused silica and metal tubing
  • Two-sided ceramic wafer
  • Serrated on one side and straight edged on the other
  • Ergonomic rubber handle
Specifications
Type Scoring Wafer