Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ SeCure Y Connector Kits
SDP

Catalog No. 06818145 Shop All Restek Corporation Products
Encompass_Preferred
missing translation for 'changeView'
Click to view available options
For Use With:
Fits Column with ID 0.32 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
06-818-145 Fits Column with ID 0.32 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 06-818-145 Supplier Restek™ missing translation for 'supplierNo' 20277
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Connect two analytical columns to a transfer line or guard column.

  • Fits columns with I.D. 0.32mm
  • Use standard ″Y″ Press-Tight connectors and 1/16 in. graphite ferrules
  • Reliable seal integrity - will not unexpectedly disconnect during temperature-programmed analyses
  • Open design allows visual confirmation of the seal for added confidence in the connection
  • Fit both Restek cage designs

Specifications

Type Connector
Diameter (Metric) Inner 0.32 mm
For Use With (Application) Connect Two Analytical Columns to a Transfer Line or Guard Column
For Use With Fits Column with ID 0.32 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.