Learn More
Restek™ Septum Nuts for use with Agilent GCs
SDP
Catalog No. 06711512 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Autosampler and PTV (for 23-Gauge Needles)
Manual Injection (for 26-Gauge Needles)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
06-711-512 Autosampler and PTV (for 23-Gauge Needles)
06-710-861 Manual Injection (for 26-Gauge Needles)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-711-512 Supplier Restek™ Supplier No. 21309
For use with Agilent 5890, 6850, 6890, 7890 split/splitless injectors.
  • Needle guide allows easy penetration and prevents septum coring
  • Made of high-quality stainless steel
  • Manual injection septum nut allows use of 26-gauge needles for on-column injections
  • Autosampler and PTV septum nut allows use of 23-gauge needles
Specifications
For Use With (Equipment) Autosampler and PTV (for 23-Gauge Needles)
Type Septum Nut