Learn More
Restek™ Septum Puller
SDP
Catalog No. 06710501 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Removes Septa, O-ring, Ferrules
Catalog No. For Use With (Application)
06-710-501 Removes Septa, O-ring, Ferrules
1 options
Catalog No. 06-710-501 Supplier Restek™ Supplier No. 20117
Features one hooked end and one pointed end
  • Hooked end for removing septa and O-rings
  • Pointed end works well for removing stuck ferrules or debris
Specifications
Type Puller
For Use With (Application) Removes Septa, O-ring, Ferrules