Learn More
Restek™ Shortix Capillary GC Column Cutter
SDP
Catalog No. 06712134 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Capillary GC Column Cutter
Maintenance Kit
For Use With (Equipment):
0.25 to 0.53mm I.D. Tubing
Shortix Capillary GC Column Cutter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
06-712-134 Maintenance Kit Shortix Capillary GC Column Cutter
06-712-133 Capillary GC Column Cutter 0.25 to 0.53mm I.D. Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-712-134 Supplier Restek™ Supplier No. 23027
Consistently make precise, clean and square cuts with a diamond blade
  • Built-in magnifier to verify square cut
  • Use with 0.25 mm ID to 0.53 mm ID tubing (0.78 mm OD maximum)
Specifications
Type Maintenance Kit
Includes Diamond cutting wheel, O-rings, tool to open the column cutter
For Use With (Application) Gas Chromatography
For Use With (Equipment) Shortix Capillary GC Column Cutter