Learn More
Restek™ Siltek™ Press-Tight™ Connectors
SDP
Catalog No. 06710638 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Presstight Connector
Packaging Quantity:
25/Pk.
Catalog No. Type Packaging Quantity
06-710-638 Presstight Connector 25/Pk.
1 options
Catalog No. 06-710-638 Supplier Restek™ Supplier No. 20449
Siltek deactivation for an inert pathway that maintains sample integrity
  • Made from inert fused silica
  • For use in connecting guard columns to analytical columns
  • Fit Restek column outer diameters from 0.33 to 0.74mm (0.1 to 0.53mm inside diameter)
  • Angled Press-Tight connector approximates the curvature of a capillary column to reduce strain on column-end connections
Specifications
Type Presstight Connector
Packaging Quantity 25/Pk.
Material Siltek