Learn More
Restek™ Siltek Septum-Purged Packed Column Port Weldment
SDP
Catalog No. 06711729 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
Each
Catalog No. Packaging Quantity
06-711-729 Each
1 options
Catalog No. 06-711-729 Supplier Restek™ Supplier No. 21691
For use with Agilent 5890 GCs.
  • Siltek treatment makes weldment inert and eliminates adsorption of sensitive compounds
  • 1 Each
Order Info
Order Viton O-rings and appropriate septa separately.
Specifications
Type Siltek Septum Packed Purge Port Weldment
Packaging Quantity Each
For Use With Agilent 5890 GCs