Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ SPI Liners for Varian, With Buffer
SDP

Catalog No. 06711147 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25/Pk.
5/Pk.
Each
Deactivation:
Intermediate Polarity (IP)
Wool
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Deactivation
06-711-147 Each Intermediate Polarity (IP)
06-718-095 5/Pk. Intermediate Polarity (IP)
06-718-104 25/Pk. Intermediate Polarity (IP)
06-718-102 5/Pk. Wool
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-711-147 Supplier Restek™ Supplier No. 20850

For Bruker/Varian GCs equipped with SPI inlets.

  • Geometry: SPI with Buffer
  • Material: Borosilicate Glass
  • Deactivation: Intermediate Polarity (IP)
  • Liner ID: 2.4mm
  • Length: 54mm
  • OD: 4.6mm

Specifications

Length (Metric) 54 mm
Product Type SPI Liner
Includes Buffer
Deactivation Intermediate Polarity (IP)
Geometry SPI with Buffer
Quantity Each
Diameter (Metric) Inner 2.4 mm
Diameter (Metric) Outer 4.6 mm
For Use With (Equipment) Bruker/Varian GCs Equipped with SPI Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.