Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Glass Liner
SDP

Catalog No. 06717806 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packing Material:
Base Deactivated
Base Deactivated Wool
CarboFrit
None
Product Type:
Cup Splitter Inlet Liner
IP Cup Splitter Inlet Liner
Quantity:
5/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packing Material Product Type Quantity
06-717-806 None IP Cup Splitter Inlet Liner 5/Pk.
06-717-810 CarboFrit IP Cup Splitter Inlet Liner 5/Pk.
06-717-811 Base Deactivated Cup Splitter Inlet Liner 5/Pk.
06-717-812 Base Deactivated Wool Cup Splitter Inlet Liner 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-717-806 Supplier Restek™ Supplier No. 20710
May include imposed supplier surcharges.
Add to Cart
Add to Cart

For high or low molecular weight compounds

  • Increased sample residence time in the liner for homogeneous vaporization
  • 4mm I.D.
  • Available with or without packing

Specifications

Packing Material None
Product Type IP Cup Splitter Inlet Liner
Length (Metric) 7.85 cm
Quantity 5/Pk.
Diameter (Metric) Inner 0.4 cm
Diameter (Metric) Outer 0.63 cm
For Use With (Equipment) Agilent GCs Equipped with Split/Splitless Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.