Learn More
Restek™ Split/Splitless Injection Port
SDP
Catalog No. 06719102 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
06-719-102 Each
1 options
Catalog No. 06-719-102 Supplier Restek™ Supplier No. 22670
For Agilent 6850/6890 GCs.
  • Manufactured from high-quality stainless steel
  • Siltek treatment passivates the metal surface to ensure an inert pathway for the sample
Specifications
Product Type Split/Splitless Injection Port
Quantity Each
For Use With (Equipment) Agilent 6890/6850 GCs, Large Canister Type Filter
Liner Material Stainless Steel