Learn More
Restek™ Stainless Steel Jet Reamers
SDP
Catalog No. 06717590 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
06-717-590 Stainless Steel
1 options
Catalog No. 06-717-590 Supplier Restek™ Supplier No. 20113
Clean detector jets for removing stuck ferrules from other small orifices
Serrated design is optimal for removing silica deposits and other contaminants.
Specifications
Type Reamer
Material Stainless Steel