Learn More
Restek™ Stainless Steel Tube Brushes/Surface Brush Set
SDP
Catalog No. 06717589 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
Set of 4
Catalog No. Packaging Quantity
06-717-589 Set of 4
1 options
Catalog No. 06-717-589 Supplier Restek™ Supplier No. 20112
Use stainless steel tube brushes for cleaning dirty collectors, injector ports, and detector ports.
  • Use surface brush to remove residue that builds up on metal detector jets and electronic contacts
  • Set of four brushes - three tube brushes (3/8 in., 3/16 in., and 1/4 in.) and one surface brush
Specifications
Type Tube Brushes/Surface Brushes
For Use With (Application) Remove Residue That Builds up on Metal Detector Jets and Electronic Contacts and for Cleaning Dirty Collectors, Injector Ports, and Detector Ports.
Packaging Quantity Set of 4
Material Stainless Steel