Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Straight Inlet Liners for Splitless Injection
SDP

Catalog No. 06711899 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5/Pk.
Diameter (Metric) Inner:
0.2 cm
0.4 cm
Includes:
Glass Splitless
Glass Splitless with Wool
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (Metric) Inner Includes
06-711-899 5/Pk. 0.4 cm Glass Splitless with Wool
06-717-822 5/Pk. 0.2 cm Glass Splitless
06-717-904 5/Pk. 0.4 cm Glass Splitless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-711-899 Supplier Restek™ Supplier No. 22401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition

  • Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
  • Compatible with Agilent™/Finnigan™ columns
  • Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples; also, packing in wool is recommended to aid in vaporization of high molecular weight compounds and to minimize discrimination
  • O.D. × Length: 6.5 × 78.5mm

Specifications

Product Type IP Splitless Straight Inlet Liner
Length (Metric) 7.85 cm
Packing Material Deactivated Wool
Includes Glass Splitless with Wool
Quantity 5/Pk.
Diameter (Metric) Inner 0.4 cm
Diameter (Metric) Outer 0.65 cm
For Use With (Equipment) Agilent GCs Equipped with Split/Splitless Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.