Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ TO-14A Aromatics Mix
SDP

Disposable and does not require rental or demurrage fees

$2555.00 - $3366.00

Specifications

Components Benzene, Chlorobenzene, m-Dichlorobenzene, o-Dichlorobenzene, p-Dichlorobenzene, Ethyl Benzene, Styrene, Toluene, 1,2,4-Trichlorobenzene, 1,2,4-Trimethylbenzene, 1,3,5-Trimethylbenzene, m-Xylene, o-Xylene, p-Xylene in Nitrogen
Volume (Metric) Gas 104 L
Pressure 1800 psi
Weight (English) 1.5 lb.
Volume (English) Gas 27.47 gal.
View More Specs

Includes: Certificate of Analysis and unique serial number

Products 2
Catalog Number Mfr. No. Concentration Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Price Quantity & Availability  
06-802-377
SDP
missing translation for 'viewDocuments'
Restek™
34404
1 ppmv
Each for $2,555.00
Only null left
Add to cart
 
06-802-388
SDP
missing translation for 'viewDocuments'
Restek™
34423
100 ppbv
Each for $3,366.00
Only null left
Add to cart
 
Description

Description

Air-monitoring ga calibration standards are provided by Spectra/Linde

  • Minimum 12-month stability on all cylinders
  • Aluminum construction
  • Size: 8 x 24cm
  • Volume/Pressure: 104L of gas at 1800 psi
  • CGA-180 outlet fitting
  • Weight: 1.5 lb. (0.7kg)

Compounds

  • Benzene
  • Chlorobenzene
  • m-Dichlorobenzene
  • o-Dichlorobenzene
  • p-Dichlorobenzene
  • Ethyl benzene
  • Styrene
  • Toluene
  • 1,2,4-Trichlorobenzene
  • 1,2,4-Trimethylbenzene
  • 1,3,5-Trimethylbenzene
  • m-Xylene
  • o-Xylene
  • p-Xylene

Order Info

Gas standards are subject to hazardous materials shipping fees by most freight carriers. All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements. Drop-shipped to provide the freshest standard possible.

Specifications

Specifications

Benzene, Chlorobenzene, m-Dichlorobenzene, o-Dichlorobenzene, p-Dichlorobenzene, Ethyl Benzene, Styrene, Toluene, 1,2,4-Trichlorobenzene, 1,2,4-Trimethylbenzene, 1,3,5-Trimethylbenzene, m-Xylene, o-Xylene, p-Xylene in Nitrogen
1800 psi
27.47 gal.
TO-14A Aromatics Mix
104 L
1.5 lb.
0.7 kg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.