Description
- Available in brass and stainless steel
- Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Note:
Nuts and ferrules are not treated unless requested. Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.
Specifications
Specifications
| For Use With (Equipment) | GC Equipment |
| Material | Brass |
| Type | Tee Fitting |
| Diameter (English) Inside | 0.25 in. |
| Quantity | 2/Pk. |