Learn More
Restek™ Tube and Pipe Fittings for GC Equipment
SDP
Catalog No. 06718743 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Nut and Ferrule Set
Tee Fitting
Diameter (English) Inside:
0.25 in.
Material:
Brass
Stainless Steel
Quantity:
2/Pk.
5/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Diameter (English) Inside Material Quantity
06-718-743 Nut and Ferrule Set 0.25 in. Stainless Steel 5/Pk.
06-718-708 Tee Fitting 0.25 in. Brass 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-718-743 Supplier Restek™ Supplier No. 23161
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Available in brass and stainless steel
  • Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Note:
Nuts and ferrules are not treated unless requested. Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.
Specifications
Type Nut and Ferrule Set
Material Stainless Steel
For Use With (Equipment) GC Equipment
Diameter (English) Inside 0.25 in.
Quantity 5/Pk.
Includes Nuts, Back Ferrules, and Front Ferrules