Learn More
Restek™ Tubing Bender
SDP
Catalog No. 06712118 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Catalog No. Packaging Quantity
06-712-118 1 Ea.
1 options
Catalog No. 06-712-118 Supplier Restek™ Supplier No. 23009
Assists in making accurate left-hand, right-hand or offset bends
Triple-head tool bends 3.18, 4.76 and 6.35mm (1/8, 3/16 and 1/4 in.) tubing
Specifications
Type Tubing Bender
Packaging Quantity 1 Ea.