Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Uniliner™ Inlet Liners: Cyclo
SDP

Catalog No. 06710588 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric) Inner:
4 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
06-710-588 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-710-588 Supplier Restek™ Supplier No. 22270

missing translation for 'validMainframeMsg'

For Agilent GCs equipped with split/splitless inlets.

Cyclo Liners feature a glass spiral that provides an excellent vaporization surface for high and low molecular weight samples. Provides up to five times as many dirty sample injections as liners packed with wool. Suitable for trace, dirty, high MW, active samples, high recovery and linearity.

Order Info

Liners with a Siltek–deactivated surface offer high temperature stability, durability, and low bleed for minimized breakdown and better sample recovery. O.D. x Length: 6.3 x 78.5mm.

†Nominal I.D. at syringe needle expulsion point.

Specifications

Product Type Sleeve
Diameter (Metric) Inner 4 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.