Learn More
Restek™ Universal Press-Tight™ Connectors: Universal Y, Deactivated
SDP
Catalog No. 06710613 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
3 Pack
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
06-710-613 Each
06-717-755 3 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-710-613 Supplier Restek™ Supplier No. 20405261
Add to Cart
Add to Cart
Connect analytical columns, guard columns, transfer lines or detectors
  • Fit column outer diameters 0.33 to 0.74mm (Restek inside diameters 0.1 to 0.53mm)
  • Deactivated Press-Tight connectors for better recovery of polar and nonpolar compounds
  • Angle approximates the curvature of a capillary column, reducing the strain on column-end connections
Specifications
Type Connector
Packaging Quantity Each