Learn More
Restek™ Universal Press-Tight™ Connectors: Y-Connector, Deactivated
SDP
Catalog No. 06717788 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Deactivated Y Connector
Catalog No. Type
06-717-788 Deactivated Y Connector
1 options
Catalog No. 06-717-788 Supplier Restek™ Supplier No. 20404261
Connect analytical columns, guard columns, transfer lines or detectors
  • Fit column outer diameters 0.33 to 0.74mm (Restek inside diameters 0.1 to 0.53mm)
  • Deactivated Press-Tight connectors for better recovery of polar and nonpolar compounds
  • Angle approximates the curvature of a capillary column, reducing the strain on column-end connections
Specifications
Type Deactivated Y Connector