Learn More
Restek™ Valco Ferrules and Nuts
SDP
Catalog No. 06717670 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Type:
Ferrule
Nut
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-717-670 Nut
06-717-669 Ferrule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-717-670 Supplier Restek™ Supplier No. 20287
For Valco connectors.
  • Stainless-steel
  • Size: 0.0625 in.
Specifications
Material Stainless Steel
Diameter (English) Inside 0.0625 in.
For Use With (Equipment) Restek Chromatoraphy Systems
Type Nut
Quantity 10 Pack