Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - BamH I Blue/White Cloning Qualified (E)

Catalog No. PRR6021 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
2500 U
Concentration:
10 U/μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PR-R6021 2500 U 10 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PR-R6021 Supplier Promega Supplier No. R6021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Capable of digesting DNA in 15 minutes or less. Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications. (G/GATC C C CTAG\G)

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 10 U/μL
Components 1 x 1mL 10X Buffer E, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2500U BamHI, 1 x 0.25mL Multi-Core™ 10X Buffer
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 5mM MgCl2, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 2500 U
Cut Site G.GATCC CCTAG.G
Product Type BamHI
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.