Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Not I Blue/White Cloning and Genome Qualified (D)

Catalog No. PRR6435 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 U
200 U
Concentration:
10 U/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PR-R6435 1000 U 10 U/μL
PR-R6431 200 U 10 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-R6435 Supplier Promega Supplier No. R6435
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capable of digesting DNA in 15 minutes or less. (GC/GGCC GC CG CCGG\CG)

  • Capable of digesting DNA in 15 minutes or less
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications
  • High concentration product no longer available

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 10 U/μL
Components 1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U NotI, 1 x 0.25mL Multi-Core™ 10X Buffer
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Capable of digesting DNA in <15 min.
Storage Buffer 10mM Tris HCl (pH 7.4), 500mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.1% Triton™ X-100, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 1000 U
Cut Site GC.GGCCGC CGCCGG.CG
Product Type NotI
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.