Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ret Antibody (RET/2599) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308578
Description
Ret Monoclonal specifically detects Ret in Human samples. It is validated for ELISA, Protein Array.Specifications
Ret | |
Monoclonal | |
1 mg/mL | |
ELISA : 2-4 ug/mL, Protein Array | |
CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary) | |
100 μg | |
Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
5979 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG2b κ |
ELISA, Peptide Array | |
RET/2599 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction