Learn More
Promega RET Kinase Enzyme System, 1mg
Catalog No. PRV3762 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3762 1 mg
1 options
Catalog No. PRV3762 Supplier Promega Supplier No. V3762
Add to Cart
Add to Cart
Request a Quote
Recombinant human RET (amino acids 658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RET gene codes for a transmembrane tyrosine kinase.
  • Easily Screen and Profile RET Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C