Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RET Monoclonal Antibody (OTI21H4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA805811

Description
RET is a member of the cadherin superfamily, encodes one of the receptor tyrosine kinases which are cell-surface molecules that transduce signals for cell growth and differentiation. RET plays a crucial role in neural crest development, can undergo oncogenic activation in vivo and in vitro by cytogenetic rearrangement. Mutations in the RET gene are associated with the disorders multiple endocrine neoplasia, type IIA, multiple endocrine neoplasia, type IIB, Hirschsprung disease, and medullary thyroid carcinoma. Two transcript variants encoding different isoforms have been found for the RET gene. Additional transcript variants have been described for RET but their biological validity has not been confirmed.Specifications
RET | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P07949 | |
RET | |
Human recombit protein fragment corresponding to amino acids 713-1017 of human RET produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI21H4 | |
Unconjugated | |
RET | |
CDHF12, CDHR16, HSCR1, MEN2A, MEN2B, MTC1, PTC, RET-ELE1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5979 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction