Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RET Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514724
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514724 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514724 Supplier Invitrogen™ Supplier No. PA514724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RET Polyclonal Antibody for Western Blot, IHC (P)

The receptor tyrosine kinase RET is a member of the cadherin superfamily. RET plays a crucial role in neural crest development, and can undergo oncogenic activation by cytogenetic rearrangement. RET transduce signals for cell growth and differentiation. Mutations in the RET gene are associated with the disorders multiple endocrine neoplasia, type IIA, multiple endocrine neoplasia, type IIB, Hirschsprung disease, and medullary thyroid carcinoma. Two transcript variants encoding different isoforms have been found for the RET gene. Additional transcript variants have been described for RET but their biological validity has not been confirmed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RET
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RET
Gene Accession No. P07949
Gene Alias Cadherin family member 12; Cadherin family member 12 (CDHF12); Cadherin related family member 16 (CDHR16); cadherin-related family member 16; CDHF12; CDHR16; CG1061; CG14396; CG14396-PA; CG14396-PB; CG14396-PC; c-Ret; CUX1/RET fusion; Dmel\CG14396; Dmel_CG14396; DmHD-59; DmRet; Dret; D-ret; EC 2.7.10.1; ELKS; Extracellular cell-membrane anchored RET cadherin 120 kDa fragment; HD-59; HSCR1; Hydroxyaryl protein kinase; hydroxyaryl-protein kinase; kinase Ret; MEN2; MEN2A; MEN2B; MTC1; Multiple endocrine neoplasia and medullary thyroid carcinoma 1; Oncogene RET; OTTHUMP00000216967; proto-oncogene c-Ret; Proto-oncogene tyrosine-protein kinase receptor Ret; PTC; rearranged during transfection; receptor tyrosine kinase; ret; RET ELE1; Ret gene for receptor tyrosin; Ret oncogene; ret proto-oncogene; ret proto-oncogene (multiple endocrine neoplasia and medullary thyroid carcinoma 1, Hirschsprung disease); Ret proto-oncogene (multiple endocrine neoplasia MEN2A MEN2B and medullary thyroid carcinoma 1 Hirschsprung disease); RET receptor tyrosine kinase; RET transforming sequence; RET51; RET9; RET-ELE1; Reto; Ret-PA; Ret-PB; Ret-PC; Soluble RET kinase fragment
Gene Symbols RET
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1012-1042 amino acids from the C-terminal region of human RET.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.