Learn More
Reticulon-3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302859AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.Specifications
Reticulon-3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95197 | |
RTN3 | |
Between 550 and 600 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RTN3 | |
HAP; ASYIP; NSPL2; NSPLII; RTN3-A1; RTN3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10313 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.