Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinal Dehydrogenase 1/ALDH1A1, Mouse anti-Human, Rat, Clone: 16B4.2, Millipore Sigma™

Catalog No. MABD634100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD634100 100 μg
MABD63425UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABD634100 Supplier MilliporeSigma Supplier No. MABD634100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Retinal Dehydrogenase 1/ALDH1A1, clone 16B4.2, Cat. No. MABD634, is a mouse monoclonal antibody that detects Retinal Dehydrogenase 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Retinal dehydrogenase 1 (UniProt: P00352; also known as EC: 1.2.1.36, RALDH 1, RalDH1, ALDH-E1, ALHDII, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic) is encoded by the ALDH1A (also known as ALDC, ALDH1, PUMB1) gene (Gene ID: 216) in human. Retinal dehydrogenase 1 is a ubiquitously distributed cytosolic enzyme that serves as a detoxification enzyme and also generates retinoic acid. It catalyzes the oxidation of retinal to retinoic acid that is essential for differentiation, development, and survival of dopaminergic neurons. It can also play a role in cellular defense against oxidative stress by oxidizing 4-hydroxynonenal (4-HNE) produced during lipid peroxidation. It is also involved in several other biological processes, such as stem cell maintenance, UV-radiation resistance, melanogenesis, and spermatogenesis, by catalyzing the conversion of specific aldehydes to their corresponding carboxylic acids. Retinal dehydrogenase 1 contains multiple NAD binding sites. (Ref.: Paval, D., et al. (2017). Clin. Psychopharmacol. Neurosci. 5(3); 229-236).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Retinal Dehydrogenase 1/ALDH1A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 16B4.2
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Retinal Dehydrogenase 1/ALDH1A1 in human liver and rat liver tissue sections.
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Retinal dehydrogenase 1;EC: 1.2.1.36;RALDH 1;RalDH1;ALDH-E1;ALHDII;Aldehyde dehydrogenase family 1 member A1;Aldehyde dehydrogenase;cytosolic
Gene Symbols ALDH1A;ALDC;ALDH1;PUMB1
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human retinal dehydrogenase 1.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000680
Test Specificity Clone 16B4.2 detects Retinal dehydrogenase 1/ALDH1A1. it targets an epitope with in 18 ammo acids from the C-terminal region.
Target Species Human, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.