Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Retinoblastoma binding protein 1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126616AF488
Description
Retinoblastoma binding protein 1 Polyclonal antibody specifically detects Retinoblastoma binding protein 1 in Human samples. It is validated for Western BlotSpecifications
Retinoblastoma binding protein 1 | |
Polyclonal | |
Western Blot | |
AT rich interactive domain 4A (RBP1-like), RBBP-1, RBP-1, RBP1ARID domain-containing protein 4A, retinoblastoma binding protein 1, Retinoblastoma-binding protein 1RBBP1AT-rich interactive domain-containing protein 4A | |
The immunogen recognized by this antibody maps to a region between residue 900 and 950 of human AT-rich interactive domain-containing protein 4A using the numbering given in entry NP_002883.3 (GeneID 5926). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
5926 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction