Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Retinoic acid
GREENER_CHOICESDP

Catalog No. 069550 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-955-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-955-0 Supplier R&D Systems™ Supplier No. 0695/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Endogenous retinoic acid receptor agonist

Retinoic acid is an endogenous agonist for retinoic acid receptors (IC50 = 14 nM for RARα, RARβ and RARγ receptors). Also promotes differentiation of mouse embryonic stem cells (ESCs) into adipocytes, neurons and glia in vitro. Proposed ligand of RORβ (Kd = 280 nM). Activates autophagy.

For more information about how Retinoic acid may be used, see our protocols: Generation of β cells from hPSCs, Cultivating Cerebral Organoids .

Chemical Identifiers

CAS 302-79-4
Molecular Formula C20H28O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 300.44
Purity >99%
Molecular Weight (g/mol) 300.44
Chemical Name or Material Retinoic acid
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.