Learn More
Retinoic acid, 97%
Description
Retinoic acid is used in dermatology drugs. It is used in the treatment of psoriasis, ichthyosis, follicular keratosis, acne, lichen planus, verrucous epidermal nevus and lichen psoriasis. It is also employed in promoting the proliferation and differentiation of epithelial cells. In addition to this, it exhibits some preventive effect on skin cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders
Applications
• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only

Specifications
Specifications
Melting Point | 180°C to 182°C |
Quantity | 0.5 g |
Beilstein | 2057223 |
Sensitivity | Light sensitive |
Merck Index | 14,8165 |
Solubility Information | Soluble in ethanol,acetate,dimethyl formamide,dimethyl sulfoxide,chloroform,methanol,acetone and dichloromethane. Insoluble in water and ether. |
IUPAC Name | (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid |
Formula Weight | 300.44 |
Percent Purity | 98% |
Physical Form | Crystalline |
Show More |
Safety and Handling
Hazard Category | H302+H312-H315-H319-H335-H361 |
Hazard Statement | GHS H Statement H361-H302 Suspected of damaging fertility or the unborn child. Harmful if swallowed. |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 206-129-0 |
RTECSNumber | VH6475000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.