Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Retinol Binding Protein RBP Antibody (OTI2H3), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273852MFV610
Description
Retinol Binding Protein RBP Monoclonal antibody specifically detects Retinol Binding Protein RBP in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Retinol Binding Protein RBP | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI2H3 | |
Flow Cytometry | |
C, Cellular retinol-binding protein, CRABP-I, CRBP1CRBP-I, CRBPCellular retinol-binding protein I, CRBPI, RBPC, retinol binding protein 1, cellular, retinol-binding protein 1, retinol-binding protein 1, cellular | |
Full length human recombinant protein of human RBP1 (NP_002890) produced in HEK293T cell. | |
0.1 mL | |
Cancer, Diabetes Research, Lipid and Metabolism | |
5947 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction