Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinol Binding Protein RBP Antibody (RBP/872) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP248017B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP248017B 0.2 mg
NBP248017A 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP248017B Supplier Novus Biologicals Supplier No. NBP2480170.2MG

Mouse Monoclonal Antibody

Retinol Binding Protein RBP Monoclonal antibody specifically detects Antigen in Goat, Human, Monkey, Mouse, Rabbit, Rat samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen Retinol Binding Protein RBP
Applications Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone RBP/872
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1 - 2 ug/ml, Immunofluorescence : 1 - 2 ug/ml
Formulation PBS with No Preservative
Gene Alias C, Cellular retinol-binding protein, CRABP-I, CRBP1CRBP-I, CRBPCellular retinol-binding protein I, CRBPI, RBPC, retinol binding protein 1, cellular, retinol-binding protein 1, retinol-binding protein 1, cellular
Gene Symbols RBP1
Host Species Mouse
Immunogen Recombinant human retinol binding protein-1 (RBP1)
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5947
Test Specificity Recognizes a protein of 21kDa-25kDa, identified as retinol binding protein-1 (RBP1). This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin, which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post-transnationally and results in defective delivery and supply to the epidermal cells.
Target Species Human, Mouse, Rat, Goat, Monkey, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.