Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Retinol Binding Protein RBP Antibody (RBP/872), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2450150.1MG
Description
Ensure accurate, reproducible results in Immunohistochemistry (Paraffin), Immunofluorescence
Retinol Binding Protein RBP Monoclonal specifically detects Retinol Binding Protein RBP in Human, Mouse, Rat, Goat, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Retinol Binding Protein RBP | |
Monoclonal | |
0.2mg/mL | |
Immunohistochemistry-Paraffin 1 - 2 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
P02753, P09455, P50120 | |
RBP1 | |
Recombinant human retinol binding protein-1 (RBP1) | |
0.1 mg | |
Primary | |
Recognizes a protein of 21kDa-25kDa, identified as retinol binding protein-1 (RBP1). This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin, which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post-transnationally and results in defective delivery and supply to the epidermal cells. | |
Store at 4C. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Immunofluorescence | |
RBP/872 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
C, Cellular retinol-binding protein, CRABP-I, CRBP1CRBP-I, CRBPCellular retinol-binding protein I, CRBPI, RBPC, retinol binding protein 1, cellular, retinol-binding protein 1, retinol-binding protein 1, cellular | |
Mouse | |
Protein A or G purified | |
RUO | |
5947 | |
Human, Mouse, Rat, Goat, Monkey, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction