Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinol, MP Biomedicals™

Catalog No. icn19026825
Click to view available options
Quantity:
25 mg
100 mg
250 mg
500 mg

Chemical Identifiers

CAS 68-26-8
Molecular Formula C20H30O
Molecular Weight (g/mol) 286.459
MDL Number MFCD00001552
InChI Key FPIPGXGPPPQFEQ-OVSJKPMPSA-N
Synonym retinol, vitamin a, all-trans-retinol, vitamin a1, alphalin, axerophthol, vitamin a alcohol, oleovitamin a, chocola a, alphasterol
PubChem CID 445354
ChEBI CHEBI:17336
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraen-1-ol
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCO)C)C

Specifications

Color Orange
Quantity 25 mg
Formula Weight 286.5
Physical Form Powder
Chemical Name or Material Retinol, Molecular biology
EINECSNumber 200-683-7
Recommended Storage Store at 0°C under nitrogen, protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.