Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinoschisin Mouse anti-Human, Mouse, Unconjugated, Clone: 3R10, MilliporeSigma™

Catalog No. MABN243425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2434-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2434-25 Supplier MilliporeSigma Supplier No. MABN243425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Retinoschisin, clone 3R10, Cat. No. MABN2434, is a highly specific mouse monoclonal antibody that targets Retinoschisin and has been tested in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Retinoschisin
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3R10
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias X-linked juvenile retinoschisis protein;RS1-3R10
Gene Symbols RS1;XLRS1
Host Species Mouse
Immunogen GST-tagged fusion protein containing a the LSSTEDEGEDPWYQKAC peptide fragment corresponding to amino acids 22-39 from the N-terminal region of human Retinoschisin precusor protein.
Molecular Weight of Antigen ∽23 kDa observed; 25.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 3R10 detects Retinoschisin in murine retinal cells. It targets an epitope within 18 amino acids from the N-terminal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.