Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Retractable Badge Holder

Catalog No. 19103139 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Gray
Red
White
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-139 Red
19-103-142 Black
19-103-141 Gray
19-103-140 White
19-103-138 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-103-139 Supplier Brady™ Supplier No. 95072
Add to Cart
Add to Cart

Stop losing ID cards. The Brady™ Retractable Badge Holders keep your ID, access or security card near, while allowing the range to scan or swipe as needed.

  • ID tag
  • Lifetime guarantee
  • Ready-to-roll, full factory assembly

  • Ideal alternative to bull dog clips which may damage clothing
  • Clip attaches to belt or pocket while the heavy-duty nylon cable with fastener attaches to badge

Specifications

Color Red
For Use With (Equipment) ID badges, access and security cards
Includes 1/Each
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.