Learn More
Description
- ID tag
- Lifetime guarantee
- Ready-to-roll, full factory assembly
- Ideal alternative to bull dog clips which may damage clothing
- Clip attaches to belt or pocket while the heavy-duty nylon cable with fastener attaches to badge
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | ID badges, access and security cards |
| Includes | 1/Each |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.