Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RETSAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565443
Description
Immunogen sequence: SSPNPFSEDVK RPPAPLVTDK EARKKVLKQA FSANQVPEKL DVVVIGSGFG GLAAAAILAK AGKRVLVLEQ HTKAGGCCHT FGKNGLEFDT GIHYIGRMEE GSIGRFILDQ ITEGQLDWAP LSSPFDIMVL EGPNGRKEYP M Highest antigen sequence identity to the following orthologs - mouse 88%, rat 89%.
Retinol saturase carrying out the saturation of the 13-14 double bond of all-trans-retinol to produce all-trans-13,14-dihydroretinol. Has activity toward all-trans-retinol as substrate. Does not use all-trans-retinoic acid nor 9-cis, 11-cis or 13-cis-retinol isomers as substrates. May play a role in the metabolism of vitamin A.
Specifications
RETSAT | |
Polyclonal | |
Unconjugated | |
RETSAT | |
All-trans-13,14-dihydroretinol saturase; all-trans-retinol 13,14-reductase; PPAR-alpha-regulated and starvation-induced gene protein; PPSIG; retinol saturase; retinol saturase (all-trans-retinol 13,14-reductase); RETSAT; UNQ439/PRO872 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54884 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.40 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NUM9 | |
RETSAT | |
Recombinant Human RETSAT. Recombinant protein control fragment (Product #RP-104822). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction