Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RETSAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552491 Supplier Invitrogen™ Supplier No. PA552491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVSVKKGHEL VNIYCPIVVS NAGLFNTYEH LLPGNARCLP GVKQQLGTVR PGLGMTSVFI CLRGTKEDLH LPSTNYYVYY DTDMDQAMER YVSMPREEAA EHIPLLFFAF PSAKDPTWE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.

Retinol saturase carrying out the saturation of the 13-14 double bond of all-trans-retinol to produce all-trans-13,14-dihydroretinol. Has activity toward all-trans-retinol as substrate. Does not use all-trans-retinoic acid nor 9-cis, 11-cis or 13-cis-retinol isomers as substrates. May play a role in the metabolism of vitamin A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RETSAT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RETSAT
Gene Accession No. Q6NUM9
Gene Alias All-trans-13,14-dihydroretinol saturase; all-trans-retinol 13,14-reductase; PPAR-alpha-regulated and starvation-induced gene protein; PPSIG; retinol saturase; retinol saturase (all-trans-retinol 13,14-reductase); RETSAT; UNQ439/PRO872
Gene Symbols RETSAT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RETSAT. Recombinant protein control fragment (Product #RP-89961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54884
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.