Learn More
RETSCH Knife Mill Grindomix GM300 Accessories: Grinding Containers
Catalog No. 08418329 Shop All RETSCH Inc. Products
Change view
Click to view available options
Product Type:
Grinding Container
Grinding Container with Knife Holder
Material:
Plastic
Stainless Steel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material
08-418-329 Grinding Container Stainless Steel
08-418-326 Grinding Container with Knife Holder Plastic
08-418-327 Grinding Container Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 08-418-329 Supplier RETSCH Supplier No. 030450055
Add to Cart
Add to Cart
Accessories for Knife Mill Grindomix GM300.
For use with Knife Mill Grindomix GM300 (Cat. No. 08-418-325).
Specifications
Product Type Grinding Container
Capacity (Metric) 5 L
For Use With (Equipment) For Knife Mill Grindomix GM300
Material Stainless Steel