Learn More
RETSCH Knife Mill Grindomix GM300 Accessories: Stainless Steel Knife
Catalog No. 08418333 Shop All RETSCH Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Knife Mill Grindomix GM300
Catalog No. For Use With (Equipment)
08-418-333 Knife Mill Grindomix GM300
1 options
Catalog No. 08-418-333 Supplier RETSCH Supplier No. 024460030
Add to Cart
Add to Cart
Accessories for Knife Mill Grindomix GM300
For use with Knife Mill Grindomix GM300 (Cat. No. 08-418-325).
  • Non-serrated blade
  • Autoclavable
Specifications
Product Type Knife, non-serrated blade
For Use With (Equipment) Knife Mill Grindomix GM300
Material Stainless Steel