Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RETSCH Safe-Lock Reaction Vials

Catalog No. 08418530 Shop All RETSCH Inc. Products
Change view
Click to view available options
Capacity (Metric):
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
08418530 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08418530 Supplier RETSCH Supplier No. 227490001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mixer mill accessories: reaction vials

  • Capacity: 2mL
  • Sold 1000/Pk.

Specifications

Capacity (Metric) 2 mL
Volume (Metric) 2 mL
Capacity (English) 0.54 dr.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.