Learn More
United Scientific™ REUZ™ Stainless Steel Balance Spoon, 175 mm
Shop All United Scientific Supplies, Inc Products
Description
- Slim blade with long cavity and a long handle enables the spoon to be used in a variety of containers.
- Constructed of a single piece of stainless steel with a polished surface to resist contamination. Intended for use with most powders, granules, pastes and liquids.
- Sold individually.
- REUZ™ products from United Scientific promote sustainability in all laboratory environments.
Specifications
Specifications
| Product Type | Balance Spoon |
| Material | Stainless Steel |
| Length (Metric) | 175 mm |
| Autoclavable | Autoclavable |
| Shape | Spoon |