Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rev-erb beta/NR1D2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Rev-erb beta/NR1D2 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Rev-erb beta/NR1D2 Polyclonal specifically detects Rev-erb beta/NR1D2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Rev-erb beta/NR1D2 | |
Polyclonal | |
Rabbit | |
Human | |
BD73, EAR-1r, HZF2, nuclear receptor subfamily 1 group D member 2, nuclear receptor subfamily 1, group D, member 2, Orphan nuclear hormone receptor BD73, rev-erba-alpha-related receptor, rev-erb-beta, RVR, V-erbA-related protein 1-related | |
NR1D2 | |
IgG | |
Affinity Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9975 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SAMKTMMNSQFSGHLQNDTLVEHHEQTALPAQEQLRPKPQLEQENIKSSSPPSSDFAKEEVIGMVTRAHKDTFMYNQEQQENSAESMQPQRGERIPKN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title