Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rev-erb beta/NR1D2 Antibody (OTI5E10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273854
Description
Rev-erb beta/NR1D2 Monoclonal specifically detects Rev-erb beta/NR1D2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Rev-erb beta/NR1D2 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000 | |
BD73, EAR-1r, HZF2, nuclear receptor subfamily 1 group D member 2, nuclear receptor subfamily 1, group D, member 2, Orphan nuclear hormone receptor BD73, rev-erba-alpha-related receptor, rev-erb-beta, RVR, V-erbA-related protein 1-related | |
Mouse | |
64.5 kDa | |
100 μg | |
9975 | |
Human, Mouse, Rat | |
Lyophilized |
Western Blot | |
OTI5E10 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
NR1D2 | |
Human recombinant protein fragment corresponding to amino acids 1-294 of human NR1D2(NP_005117) produced in E.coli. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction