Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rev-erb beta/NR1D2 Antibody (OTI5E10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273854H
Description
Rev-erb beta/NR1D2 Monoclonal specifically detects Rev-erb beta/NR1D2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Rev-erb beta/NR1D2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NR1D2 | |
Recombinant protein fragment corresponding to amino acids 1-294 of human Rev-erb beta/NR1D2 (NP_005117) produced in E.coli. | |
0.1 mL | |
9975.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI5E10 | |
Western Blot | |
BD73, EAR-1r, HZF2, nuclear receptor subfamily 1 group D member 2, nuclear receptor subfamily 1, group D, member 2, Orphan nuclear hormone receptor BD73, rev-erba-alpha-related receptor, rev-erb-beta, RVR, V-erbA-related protein 1-related | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction