Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REV1 Monoclonal Antibody (10#), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543606
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543606 100 μL
PIMA543607 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543606 Supplier Invitrogen™ Supplier No. MA543606
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This gene encodes a protein with similarity to the S. cerevisiae mutagenesis protein Rev1. The Rev1 proteins contain a BRCT domain, which is important in protein-protein interactions. A suggested role for the human Rev1-like protein is as a scaffold that recruits DNA polymerases involved in translesion synthesis (TLS) of damaged DNA. Two alternatively spliced transcript variants that encode different proteins have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REV1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10#
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Rev1
Gene Accession No. Q9UBZ9
Gene Alias 1110027I23Rik; AIBP80; alpha integrin-binding protein 80; AU022044; DNA repair protein REV1; REV1; REV1 homolog; REV1 homolog (S. cerevisiae); REV1- like; REV1, DNA directed polymerase; REV1, polymerase (DNA directed); REV1L; rev1-like terminal deoxycytidyl transferase
Gene Symbols Rev1
Host Species Mouse
Immunogen Ser301-Lys478 (Accession Q9UBZ9), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.