Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ REV3L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564186
Description
Immunogen sequence: KLDQAYTPNF LHCKDSQQQI VCIAEQSKHS ETCSPGNTAS EESQMPNNCF VTSLRSPIKQ IAWEQKQRGF ILDMSNFKPE RVKPRSLSEA ISQTKALSQ Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 78%.
Catalytic subunit of the DNA polymerase zeta complex, an error-prone polymerase specialized in translesion DNA synthesis (TLS). Lacks an intrinsic 3'-5' exonuclease activity and thus has no proofreading function.
Specifications
REV3L | |
Polyclonal | |
Unconjugated | |
REV3L | |
DNA polymerase zeta catalytic subunit; POLZ; Protein reversionless 3-like; Rev; REV3; REV3 like, DNA directed polymerase zeta catalytic subunit; Rev-3, yeast, homolog-like (polymerase, DNA, zeta); Rev3l; REV3-like; REV3-like, catalytic subunit of DNA polymerase zeta; REV3-like, catalytic subunit of DNA polymerase zeta RAD54 like (S. cerevisiae); REV3-like, polymerase (DNA directed), zeta, catalytic subunit; Seizure-related protein 4; Sez4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5980 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60673 | |
REV3L | |
Recombinant protein corresponding to Human REV3L. Recombinant protein control fragment (Product #RP-103573). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction